siRNA Details
| Gene Symbol: Rbbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175023.3 | NP_778188.1 | 3 | 699 | ||
| GenBank mRNA | AK045635.1 | - | 3 | 444 | ||
| AK081261.1 | - | 3 | 563 | |||
| AK082686.1 | - | 1 | 41 | |||
| BC025874.1 | - | 3 | 336 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbbp6| Gene Symbol: Rbbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175023.3 | NP_778188.1 | 3 | 699 | ||
| GenBank mRNA | AK045635.1 | - | 3 | 444 | ||
| AK081261.1 | - | 3 | 563 | |||
| AK082686.1 | - | 1 | 41 | |||
| BC025874.1 | - | 3 | 336 | |||