siRNA Details
| Gene Symbol: Rbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011254.5 | NP_035384.1 | 2 | 2121 | ||
| GenBank mRNA | AK011641.1 | - | 2 | 269 | ||
| BC018254.1 | - | 2 | 248 | 3498107 | ||
| BC091751.1 | - | 2 | 265 | 4022443 | ||
| X60367.1 | - | 2 | 2120 | |||

Rbp1
Genomic Map