siRNA Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104616.1 | NP_001098086.1 | 7, 8 | 845 | ||
| NM_001104617.1 | NP_001098087.1 | 7, 8 | 1251 | |||
| NM_009041.3 | NP_033067.2 | 7, 8 | 1251 | |||
| GenBank mRNA | AK016711.1 | - | 7, 8 | 921 | ||
| AK088292.1 | - | 7, 8 | 958 | |||
| AK146423.1 | - | 7, 8 | 909 | |||
| AK156370.1 | - | 7, 8 | 937 | |||
| AK162204.1 | - | 7, 8 | 823 | |||
| AK168449.1 | - | 7, 8 | 922 | |||
| BC002041.1 | - | 7, 8 | 896 | |||
| BC026786.1 | - | 7, 8 | 924 | |||
| BC034438.1 | - | 7, 8 | 1077 | |||
| BC053417.1 | - | 7, 8 | 925 | 6707873 | ||
| X60672.1 | - | 7, 8 | 864 | |||

Rdx
Genomic Map