siRNA Details
| Gene Symbol: Rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009044.2 | NP_033070.2 | 7 | 1024 | ||
| GenBank mRNA | AK031627.1 | - | 7 | 822 | ||
| BC139447.1 | - | 7 | 964 | 8862469 | ||
| BC139448.1 | - | 7 | 964 | 8862470 | ||
| BC139770.1 | - | 7 | 966 | 40111461 | ||
| X15842.1 | - | 7 | 1146 | |||
| X60271.1 | - | 7 | 1023 | |||

Rel
Genomic Map