siRNA Details
| Gene Symbol: Sbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011321.3 | NP_035451.1 | 3 | 512 | ||
| GenBank mRNA | AK035423.1 | - | 3 | 513 | ||
| BC049605.1 | - | 3 | 512 | 6743567 | ||
| X06246.1 | - | 3 | 512 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbp| Gene Symbol: Sbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011321.3 | NP_035451.1 | 3 | 512 | ||
| GenBank mRNA | AK035423.1 | - | 3 | 513 | ||
| BC049605.1 | - | 3 | 512 | 6743567 | ||
| X06246.1 | - | 3 | 512 | |||