siRNA Details
| Gene Symbol: Zc3h7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081016.1 | NP_001074485.1 | 8 | 1109 | ||
| GenBank mRNA | AK129268.1 | - | 8 | 786 | ||
| AK147484.1 | - | 8 | 873 | |||
| AK147978.1 | - | 8 | 882 | |||
| AK157711.1 | - | 1 | 50 | |||
| AK165492.1 | - | 8 | 901 | |||
| AK171949.1 | - | 8 | 824 | |||
| BC059923.1 | - | 8 | 864 | |||

Zc3h7b
Genomic Map