siRNA Details
| Gene Symbol: Shfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009169.2 | NP_033195.1 | 3 | 293 | ||
| GenBank mRNA | AK135462.1 | - | 246 | |||
| AK160432.1 | - | 3 | 286 | |||
| AK161690.1 | - | 3 | 266 | |||
| AK167914.1 | - | 3 | 243 | |||
| BC058117.1 | - | 3 | 259 | 6412144 | ||
| U41626.1 | - | 3 | 235 | |||

Shfm1
Genomic Map