siRNA Details
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006067.4 | NP_006058.1 | 3, 4 | 623 | ||
| XM_017022867.1 | XP_016878356.1 | 3, 4 | 314 | |||
| GenBank mRNA | AF005888.1 | - | 3, 4 | 492 | ||
| AK313977.1 | - | 3, 4 | 605 | |||
| BC001472.2 | - | 3, 4 | 575 | 3138396 | ||
| BC005886.2 | - | 3, 4 | 575 | 3546377 | ||
| BC007445.1 | - | 3, 4 | 566 | 3009805 | ||
| BC020250.1 | - | 3, 4 | 575 | 4653699 | ||
| BT007132.1 | - | 3, 4 | 376 | |||

EMC8
Genomic Map