siRNA Details
| Gene Symbol: Sprr2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011468.4 | NP_035598.2 | 2 | 2603 | ||
| Gene Symbol: Sprr2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164787.1 | NP_001158259.1 | 2603 | |||

Sprr2a1
Sprr2a2
Genomic Map