siRNA Details
| Gene Symbol: Gm609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005854.2 | NP_001005854.2 | 5 | 951 | ||
| GenBank mRNA | AB853320.1 | - | 5 | 967 | ||
| AK136536.1 | - | 4 | 2791 | |||
| BC054854.1 | - | 5 | 950 | 6529072 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm609| Gene Symbol: Gm609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005854.2 | NP_001005854.2 | 5 | 951 | ||
| GenBank mRNA | AB853320.1 | - | 5 | 967 | ||
| AK136536.1 | - | 4 | 2791 | |||
| BC054854.1 | - | 5 | 950 | 6529072 | ||