siRNA Details
| Gene Symbol: Iqcj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177585.3 | NP_808253.1 | 3 | 191 | ||
| GenBank mRNA | AK052136.1 | - | 3 | 192 | ||
| BC107305.1 | - | 3 | 155 | 40053239 | ||
| BC107306.1 | - | 3 | 155 | 40053241 | ||
| Gene Symbol: Gm21949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113419.2 | NP_001106890.1 | 3 | 191 | ||
| GenBank mRNA | KJ941154.1 | - | 3 | 179 | ||

Iqcj
Genomic Map