siRNA Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081006.1 | NP_001074475.1 | 4 | 776 | ||
| NM_001177630.2 | NP_001171101.2 | 3 | 874 | |||
| NM_001177631.1 | NP_001171102.1 | 3 | 874 | |||
| NM_029895.4 | NP_084171.2 | 4 | 1087 | |||
| NM_178059.5 | NP_835160.2 | 4 | 1087 | |||
| GenBank mRNA | AB093289.2 | - | 3 | 664 | ||
| AB125594.2 | - | 4 | 1087 | |||
| AB125595.2 | - | 4 | 1087 | |||
| AK052020.1 | - | 5 | 1189 | |||
| AK052733.1 | - | 3 | 562 | |||
| AK147510.1 | - | 3 | 781 | |||
| BC050016.1 | - | 3 | 577 | |||
| BC158051.1 | - | 3 | 596 | 9007405 | ||

Etl4
Genomic Map