siRNA Details
| Gene Symbol: Zdhhc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172465.4 | NP_766053.1 | 3 | 638 | ||
| GenBank mRNA | AK032233.1 | - | 3 | 620 | ||
| AK035745.1 | - | 2 | 2061 | |||
| AK079248.1 | - | 3 | 605 | |||
| BC042618.1 | - | 3 | 497 | 4950217 | ||
| BC090832.1 | - | 3 | 456 | 6401763 | ||

Zdhhc9
Genomic Map