siRNA Details
Gene Symbol: Zfp710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145999.1 | NP_001139471.1 | 2 | 1513 | ||
NM_001146000.1 | NP_001139472.1 | 2 | 1513 | |||
NM_175433.5 | NP_780642.3 | 2 | 1596 | |||
GenBank mRNA | AK036711.1 | - | 1 | 171 | ||
AK083979.1 | - | 2 | 1438 | |||
AK089457.1 | - | 1 | 788 | |||
AK154871.1 | - | 2 | 1494 | |||
AK154953.1 | - | 2 | 1514 | |||
AK155416.1 | - | 2 | 1575 | |||
BC058640.1 | - | 2 | 1407 | 6835601 | ||
BC060641.1 | - | 2 | 1490 | 6849964 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
