siRNA Details
| Gene Symbol: Frmd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177843.1 | NP_001171314.1 | 17 | 2068 | ||
| NM_001177844.1 | NP_001171315.1 | 6 | 1196 | |||
| NM_172475.3 | NP_766063.3 | 17 | 1698 | |||
| GenBank mRNA | AB093292.1 | - | 3 | 392 | ||
| AK077327.1 | - | 6 | 1196 | |||
| AK082365.1 | - | 17 | 1644 | |||
| AK148414.1 | - | 6 | 1195 | |||
| AK171532.1 | - | 6 | 1124 | |||
| BC058672.1 | - | 17 | 1689 | 6830456 | ||
| BC082808.1 | - | 17 | 2075 | |||

Frmd4a
Genomic Map