siRNA Details
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081370.2 | NP_001074839.2 | 10 | 1686 | ||
| NM_001113373.2 | NP_001106844.2 | 15 | 2132 | |||
| GenBank mRNA | AB099695.1 | - | 15 | 1771 | ||
| AK220363.1 | - | 8 | 701 | |||

Shank2
Genomic Map