siRNA Details
| Gene Symbol: Lrch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081173.1 | NP_001074642.1 | 20, 21 | 2225 | ||
| GenBank mRNA | AK122516.1 | - | 8, 9 | 1989 | ||
| BC056180.1 | - | 14, 15 | 1179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrch2| Gene Symbol: Lrch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081173.1 | NP_001074642.1 | 20, 21 | 2225 | ||
| GenBank mRNA | AK122516.1 | - | 8, 9 | 1989 | ||
| BC056180.1 | - | 14, 15 | 1179 | |||