siRNA Details
Gene Symbol: Tbc1d31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081396.2 | NP_001074865.2 | 15 | 2346 | ||
NM_001167679.1 | NP_001161151.1 | 15 | 2346 | |||
GenBank mRNA | AB185844.1 | - | 15 | 2314 | ||
AK084542.1 | - | 14 | 2110 | |||
AK141805.1 | - | 15 | 2216 | |||
AK154264.1 | - | 15 | 2311 | |||
AK161403.1 | - | 14 | 2163 | |||
AK171024.1 | - | 15 | 2311 | |||
BC050039.1 | - | 15 | 2292 | 30016266 | ||
BC066830.1 | - | 15 | 2278 | 30463017 |
