siRNA Details
| Gene Symbol: Ighv1-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF045502.1 | - | 2 | 205 | ||
| AJ006946.1 | - | 2 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ighv1-54| Gene Symbol: Ighv1-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF045502.1 | - | 2 | 205 | ||
| AJ006946.1 | - | 2 | 205 | |||