siRNA Details
| Gene Symbol: Ubxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285928.1 | NP_001272857.1 | 4 | 930 | ||
| NM_001285929.1 | NP_001272858.1 | 3 | 819 | |||
| NM_001285930.1 | NP_001272859.1 | 3 | 683 | |||
| NM_178671.5 | NP_848786.1 | 2 | 552 | |||
| GenBank mRNA | AK045964.1 | - | 2 | 510 | ||
| AK046949.1 | - | 4 | 931 | |||
| AK077656.1 | - | 2 | 554 | |||
| AK080662.1 | - | 2 | 518 | |||
| AK133185.1 | - | 2 | 538 | |||
| AK161245.1 | - | 2 | 539 | |||
| AK161296.1 | - | 2 | 515 | |||
| BC027099.1 | - | 3 | 683 | 5041000 | ||

Ubxn10
Genomic Map