siRNA Details
| Gene Symbol: Chpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146690.1 | NP_001140162.1 | 7 | 1166 | ||
| NM_144807.3 | NP_659056.3 | 7 | 1166 | |||
| GenBank mRNA | AK007964.1 | - | 7 | 1086 | ||
| AK032497.1 | - | 7 | 1061 | |||
| AY445814.1 | - | 7 | 1061 | |||
| BE650891.1 | - | 3 | 401 | |||

Chpt1
Genomic Map