siRNA Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034858.3 | NP_001030030.2 | 15 | 2245 | ||
| GenBank mRNA | AK131686.1 | - | 11 | 1442 | ||
| AK141039.1 | - | 9 | 2577 | |||
| BC172168.1 | - | 10 | 1987 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc2| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034858.3 | NP_001030030.2 | 15 | 2245 | ||
| GenBank mRNA | AK131686.1 | - | 11 | 1442 | ||
| AK141039.1 | - | 9 | 2577 | |||
| BC172168.1 | - | 10 | 1987 | |||