siRNA Details
| Gene Symbol: Nutm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172521.1 | NP_766109.1 | 7 | 2584 | ||
| GenBank mRNA | AK077049.1 | - | 7 | 2584 | ||
| BC125332.1 | - | 7 | 2564 | 40129847 | ||
| BC125334.1 | - | 7 | 2564 | 40129849 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nutm1| Gene Symbol: Nutm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172521.1 | NP_766109.1 | 7 | 2584 | ||
| GenBank mRNA | AK077049.1 | - | 7 | 2584 | ||
| BC125332.1 | - | 7 | 2564 | 40129847 | ||
| BC125334.1 | - | 7 | 2564 | 40129849 | ||