siRNA Details
| Gene Symbol: Crygn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153076.2 | NP_694716.2 | 3 | 398 | ||
| GenBank mRNA | AF445456.1 | - | 3 | 398 | ||
| BC120913.1 | - | 3 | 398 | 40091405 | ||
| BC120914.1 | - | 2 | 387 | |||
| CF739670.1 | - | 2 | 382 | |||

Crygn
Genomic Map