siRNA Details
| Gene Symbol: 4932411E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172534.2 | NP_766122.1 | 3 | 294 | ||
| GenBank mRNA | AK029964.1 | - | 3 | 295 | ||
| BC140961.1 | - | 3 | 280 | 9055573 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 4932411E22Rik
4932411E22Rik| Gene Symbol: 4932411E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172534.2 | NP_766122.1 | 3 | 294 | ||
| GenBank mRNA | AK029964.1 | - | 3 | 295 | ||
| BC140961.1 | - | 3 | 280 | 9055573 | ||
 Back To Top
 Back To Top