siRNA Details
| Gene Symbol: Lman2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013374.1 | NP_001013392.1 | 2 | 268 | ||
| GenBank mRNA | AK142024.1 | - | 266 | |||
| AK158015.1 | - | 2 | 287 | |||
| BC027226.1 | - | 2 | 269 | 2654285 | ||
| BC046969.1 | - | 2 | 268 | 6313679 | ||

Lman2l
Genomic Map