siRNA Details
| Gene Symbol: Prr14l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194340.2 | NP_919321.2 | 10 | 5869 | ||
| GenBank mRNA | AK049285.1 | - | 3 | 201 | ||
| AY372183.1 | - | 5 | 1090 | |||
| BC059908.1 | - | 3 | 301 | |||
| BC094374.1 | - | 5 | 1416 | 30531815 | ||

Prr14l
Genomic Map