siRNA Details
| Gene Symbol: Myl6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172259.1 | NP_758463.1 | 6 | 652 | ||
| GenBank mRNA | BC037527.1 | - | 6 | 652 | 3468485 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myl6b| Gene Symbol: Myl6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172259.1 | NP_758463.1 | 6 | 652 | ||
| GenBank mRNA | BC037527.1 | - | 6 | 652 | 3468485 | |