siRNA Details
| Gene Symbol: Plekhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177503.1 | NP_001170974.1 | 13 | 2150 | ||
| GenBank mRNA | AK028383.1 | - | 13 | 2100 | ||
| AK082841.1 | - | 2 | 886 | |||
| BC137602.1 | - | 13 | 1933 | 8860621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhd1| Gene Symbol: Plekhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177503.1 | NP_001170974.1 | 13 | 2150 | ||
| GenBank mRNA | AK028383.1 | - | 13 | 2100 | ||
| AK082841.1 | - | 2 | 886 | |||
| BC137602.1 | - | 13 | 1933 | 8860621 | ||