siRNA Details
| Gene Symbol: Larp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172585.2 | NP_766173.1 | 7 | 903 | ||
| GenBank mRNA | AK042491.1 | - | 7 | 901 | ||
| AK080953.1 | - | 7 | 927 | |||
| AK160759.1 | - | 7 | 904 | |||
| AK172916.1 | - | 7 | 885 | |||
| BC059218.1 | - | 7 | 883 | |||
| BC078640.1 | - | 7 | 826 | 6506267 | ||
| BC086626.1 | - | 7 | 883 | |||

Larp4b
Genomic Map