siRNA Details
| Gene Symbol: Amph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289546.1 | NP_001276475.1 | 6 | 506 | ||
| NM_175007.2 | NP_778172.1 | 6 | 506 | |||
| GenBank mRNA | AK047144.1 | - | 6 | 460 | ||
| AK132168.1 | - | 6 | 507 | |||
| BC054718.1 | - | 6 | 486 | 5686478 | ||

Amph
Genomic Map