siRNA Details
| Gene Symbol: Tnni2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009405.2 | NP_033431.1 | 7 | 626 | ||
| GenBank mRNA | BC028515.1 | - | 8 | 664 | 1448494 | |
| J04992.1 | - | 7 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnni2| Gene Symbol: Tnni2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009405.2 | NP_033431.1 | 7 | 626 | ||
| GenBank mRNA | BC028515.1 | - | 8 | 664 | 1448494 | |
| J04992.1 | - | 7 | 626 | |||