siRNA Details
| Gene Symbol: Tpbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164792.1 | NP_001158264.1 | 2 | 1518 | ||
| NM_011627.4 | NP_035757.2 | 2 | 1627 | |||
| GenBank mRNA | AK051162.1 | - | 2 | 1598 | ||
| AK143519.1 | - | 2 | 1457 | |||
| AK165553.1 | - | 2 | 1513 | |||
| AK165597.1 | - | 2 | 1630 | |||
| BC058198.1 | - | 2 | 1486 | 5353871 | ||

Tpbg
Genomic Map