siRNA Details
| Gene Symbol: Tsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011650.3 | NP_035780.1 | 6 | 659 | ||
| GenBank mRNA | AF234179.1 | - | 6 | 659 | ||
| AK010737.1 | - | 6 | 657 | |||
| AK033003.1 | - | 3 | 1751 | |||
| AK150529.1 | - | 6 | 708 | |||
| AK151448.1 | - | 6 | 710 | |||
| AK165598.1 | - | 6 | 698 | |||
| AK201202.1 | - | 3 | 302 | |||
| AK219078.1 | - | 3 | 320 | |||
| BC004615.1 | - | 6 | 705 | 3585366 | ||
| X81464.1 | - | 6 | 659 | |||

Tsn
Genomic Map