siRNA Details
| Gene Symbol: Upk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009476.2 | NP_033502.2 | 5 | 499 | ||
| GenBank mRNA | BC104260.1 | - | 5 | 499 | 40039813 | |
| BC104261.1 | - | 5 | 499 | 40039814 | ||
| CX565093.1 | - | 5 | 709 | |||
| DQ358978.1 | - | 5 | 438 | |||
| U08030.1 | - | 5 | 497 | |||

Upk2
Genomic Map