siRNA Details
Gene Symbol: Vegfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185164.1 | NP_001172093.1 | 7 | 978 | ||
NM_011697.3 | NP_035827.1 | 7 | 1079 | |||
GenBank mRNA | AK148188.1 | - | 7 | 1078 | ||
AK210361.1 | - | 1 | 144 | |||
AK214445.1 | - | 1 | 144 | |||
BC046303.1 | - | 7 | 956 | 6492277 | ||
U43836.1 | - | 7 | 1031 |
