siRNA Details
| Gene Symbol: TRIM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199573.1 | NP_001186502.1 | 8 | 1598 | ||
| NM_006074.4 | NP_006065.2 | 8 | 1610 | |||
| GenBank mRNA | AK298715.1 | - | 9 | 1290 | ||
| AK298934.1 | - | 8 | 1164 | |||
| AK301192.1 | - | 7 | 1081 | |||
| AL360190.1 | - | 1 | 21 | |||
| AM040752.1 | - | 5 | 558 | |||
| BC022281.1 | - | 8 | 1417 | 4718024 | ||
| BC035582.1 | - | 8 | 1383 | 5583800 | ||
| HQ842635.1 | - | 7 | 1231 | |||
| X82200.1 | - | 8 | 1341 | |||

TRIM22
Genomic Map