siRNA Details
| Gene Symbol: Cpped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146067.3 | NP_666179.2 | 1 | 147 | ||
| GenBank mRNA | AK029379.1 | - | 1 | 71 | ||
| AK029442.1 | - | 1 | 170 | |||
| AK032288.1 | - | 1 | 110 | |||
| AK041329.1 | - | 1 | 120 | |||
| AK049721.1 | - | 1 | 148 | |||
| AK080876.1 | - | 1 | 147 | |||
| AK133977.1 | - | 1 | 140 | |||
| AK134718.1 | - | 1 | 148 | |||
| BC025476.1 | - | 1 | 68 | |||
| BC034347.1 | - | 1 | 127 | 2654874 | ||

Cpped1
Genomic Map