siRNA Details
| Gene Symbol: Fam43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177632.3 | NP_808300.1 | 1 | 2540 | ||
| GenBank mRNA | AK083036.1 | - | 1 | 2540 | ||
| AK137950.1 | - | 1 | 2541 | |||
| BC022623.1 | - | 1 | 1133 | |||
| BC026202.1 | - | 2 | 750 | |||
| BC099947.1 | - | 1 | 2553 | 30088416 | ||

Fam43a
Genomic Map