siRNA Details
| Gene Symbol: Cyyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144853.3 | NP_659102.1 | 1 | 313 | ||
| GenBank mRNA | AF442733.1 | - | 1 | 48 | ||
| AK158088.1 | - | 1 | 371 | |||
| AK163137.1 | - | 358 | ||||
| AK171789.1 | - | 849 | ||||
| AK171936.1 | - | 1 | 338 | |||
| AY061854.1 | - | 1 | 335 | |||
| BC099957.1 | - | 1 | 313 | 6431603 | ||
| BC125594.1 | - | 1 | 107 | 40130109 | ||
| BC125596.1 | - | 1 | 107 | 40130111 | ||

Cyyr1
Genomic Map