siRNA Details
| Gene Symbol: Fhod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289654.1 | NP_001276583.1 | 12, 13 | 2421 | ||
| NM_001289655.1 | NP_001276584.1 | 13, 14 | 2778 | |||
| NM_175276.4 | NP_780485.2 | 14, 15 | 2874 | |||
| GenBank mRNA | AB078608.1 | - | 14, 15 | 2052 | ||
| AB078609.1 | - | 12, 13 | 1599 | |||
| AB100088.1 | - | 13, 14 | 1956 | |||
| AK148245.1 | - | 12, 13 | 1564 | |||
| AK220413.1 | - | 9, 10 | 1080 | |||
| BC056371.1 | - | 4, 5 | 506 | |||
| BC138046.1 | - | 12, 13 | 1521 | 8861065 | ||

Fhod3
Genomic Map