siRNA Details
| Gene Symbol: Reep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204914.1 | NP_001191843.1 | 2, 3 | 265 | ||
| NM_144865.2 | NP_659114.2 | 2, 3 | 265 | |||
| GenBank mRNA | AY562230.1 | - | 2, 3 | 98 | ||
| BC020184.1 | - | 2, 3 | 196 | 4505679 | ||
| CX216997.1 | - | 2, 3 | 265 | |||
| EU555417.1 | - | 2, 3 | 98 | |||

Reep2
Genomic Map