siRNA Details
| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039485.4 | NP_001034574.4 | 50 | 8710 | ||
| GenBank mRNA | AK031422.1 | - | 9 | 1369 | ||
| BC026614.1 | - | 7 | 1098 | |||
| BC075677.1 | - | 11 | 1801 | |||
| BC147606.1 | - | 9 | 1263 | 9056596 | ||
| HQ215521.1 | - | 50 | 7790 | |||

Piezo2
Genomic Map