siRNA Details
| Gene Symbol: Cplx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145493.1 | NP_663468.1 | 3 | 535 | ||
| GenBank mRNA | AY264291.1 | - | 3 | 424 | ||
| BC016082.1 | - | 3 | 535 | 4501240 | ||
| BC029011.1 | - | 3 | 483 | 5357047 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cplx4| Gene Symbol: Cplx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145493.1 | NP_663468.1 | 3 | 535 | ||
| GenBank mRNA | AY264291.1 | - | 3 | 424 | ||
| BC016082.1 | - | 3 | 535 | 4501240 | ||
| BC029011.1 | - | 3 | 483 | 5357047 | ||