siRNA Details
| Gene Symbol: Cox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144874.4 | NP_659123.2 | 4 | 535 | ||
| GenBank mRNA | AK048099.1 | - | 4 | 517 | ||
| AK051792.1 | - | 4 | 433 | |||
| AK141631.1 | - | 4 | 467 | |||
| BC011509.1 | - | 4 | 495 | 3981950 | ||
| BC021498.1 | - | 4 | 455 | |||

Cox15
Genomic Map