siRNA Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081225.1 | NP_001074694.1 | 5 | 1473 | ||
| GenBank mRNA | AK031688.1 | - | 5 | 1445 | ||
| AK036700.1 | - | 3 | 1050 | |||
| AK045983.1 | - | 5 | 1429 | |||
| AK147588.1 | - | 5 | 1433 | |||
| BC150999.1 | - | 5 | 1326 | 9087912 | ||
| BC157937.1 | - | 5 | 1326 | 9088117 | ||
| BC172097.1 | - | 5 | 1326 | 9054383 | ||

Fam178a
Genomic Map