siRNA Details
| Gene Symbol: Epb4.1l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145506.4 | NP_663481.2 | 17 | 1686 | ||
| GenBank mRNA | AK034676.1 | - | 17 | 1608 | ||
| AK044959.1 | - | 17 | 1624 | |||
| AK044996.1 | - | 17 | 1632 | |||
| AK047523.1 | - | 17 | 1613 | |||
| AK173198.1 | - | 17 | 1644 | |||

Epb4.1l5
Genomic Map