siRNA Details
| Gene Symbol: Sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110791.1 | NP_001104261.1 | 4 | 732 | ||
| NM_011750.2 | NP_035880.2 | 4 | 732 | |||
| GenBank mRNA | AK133895.1 | - | 4 | 521 | ||
| AK146151.1 | - | 4 | 493 | |||
| AK147081.1 | - | 4 | 501 | |||
| AK152914.1 | - | 3 | 434 | |||
| AK154705.1 | - | 4 | 501 | |||
| AK157845.1 | - | 4 | 501 | |||
| BC009091.1 | - | 4 | 477 | 3157495 | ||
| BC055370.1 | - | 4 | 491 | 30055109 | ||
| X85802.1 | - | 4 | 489 | |||

Sf1
Genomic Map