siRNA Details
| Gene Symbol: D630003M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131021.2 | NP_001124493.1 | 11 | 2576 | ||
| NM_177657.5 | NP_808325.2 | 11 | 2576 | |||
| GenBank mRNA | AK052605.1 | - | 11 | 2575 | ||
| AK168367.1 | - | 11 | 2579 | |||
| BC137959.1 | - | 11 | 2487 | 8860978 | ||
| BC137960.1 | - | 11 | 2487 | 8860979 | ||

D630003M21Rik
Genomic Map