siRNA Details
| Gene Symbol: B930041F14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178699.4 | NP_848814.3 | 1 | 1117 | ||
| GenBank mRNA | AB041660.1 | - | 1 | 638 | ||
| AK046537.1 | - | 1 | 1120 | |||
| AK047240.1 | - | 1 | 988 | |||
| AK048841.1 | - | 1 | 962 | |||
| BC147231.1 | - | 1 | 943 | 9056274 | ||
| BC147232.1 | - | 1 | 943 | 9056277 | ||

B930041F14Rik
Genomic Map